Learn More
Selleck Chemical LLC 5-Bromoindole S5439-25mg
SDP
Catalog No. 503111526
Encompass_Preferred
5-bromoindole is an important pharmaceutical chemical intermediate and a potential inhibitor of glycogen synthase kinase 3 (GSK-3)
Catalog No. 50-311-1526 Supplier Selleck Chemical LLC Supplier No. S543925MG
5-bromoindole is an important pharmaceutical chemical intermediate and a potential inhibitor of glycogen synthase kinase 3 (GSK-3)