Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoindoline 98.0+%, TCI America™
SDP

Catalog No. B38855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38855G 5 g
B38851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38855G Supplier TCI America Supplier No. B38855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22190-33-6
Molecular Formula C8H8BrN
Molecular Weight (g/mol) 198.063
MDL Number MFCD00027410
InChI Key QEDCHCLHHGGYBT-UHFFFAOYSA-N
Synonym 5-bromoindoline, 1h-indole, 5-bromo-2,3-dihydro, 5-bromo-indoline, zlchem 28, 5-bromodihydroindole, pubchem7319, acmc-1cez6, 5-bromo-2,3-dihydro-indole, ksc497i6r
PubChem CID 3411566
IUPAC Name 5-bromo-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)Br

Specifications

Melting Point 41°C
Boiling Point 132°C
Quantity 5 g
Formula Weight 198.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindoline
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.