Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoindoxyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A0940100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0940100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0940100MG Supplier TCI America Supplier No. A0940100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17357-14-1
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
MDL Number MFCD00037933
InChI Key KFTGECHXNQBTNZ-UHFFFAOYSA-N
Synonym 3-acetoxy-5-bromoindole, 5-bromoindoxyl acetate, 1h-indol-3-ol, 5-bromo-, acetate ester, o-acetyl-5-bromoindoxyl, 1h-indol-3-ol, 5-bromo-, 3-acetate, 3-acetoxy-5-bromo-1h-indole, 5-bromoindoxyl-3-acetate, acmc-209ss9, 5-bromoindol-3-yl acetate, 5-bromo-3-indolyl acetate
PubChem CID 87075
IUPAC Name (5-bromo-1H-indol-3-yl) acetate
SMILES CC(=O)OC1=CNC2=C1C=C(C=C2)Br

Specifications

Melting Point 133°C
Color Purple-White
Quantity 100 mg
Formula Weight 254.08
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindoxyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.