Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoisatoic Anhydride 98.0+%, TCI America™
SDP

Catalog No. B36595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36595G Supplier TCI America Supplier No. B36595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4692-98-2
Molecular Formula C8H4BrNO3
Molecular Weight (g/mol) 242.03
MDL Number MFCD00016921
InChI Key DXSMYDSFWCOSFM-UHFFFAOYSA-N
Synonym 5-bromoisatoic anhydride, 6-bromo-2h-3,1-benzoxazine-2,4 1h-dione, 6-bromo-1h-benzo d 1,3 oxazine-2,4-dione, 6-bromo isatinic anhydride, 5-bromoisatoicanhydride, 6-bromo-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione, 2h-3,1-benzoxazine-2,4 1h-dione, 6-bromo, 6-bromo-1,2-dihydro-4h-3,1-benzoxazine-2,4-dione, 5-bromo-isatoic anhydride, 6-bromo-1h-benzo d 1,3-oxazaperhydroine-2,4-dione
PubChem CID 288561
IUPAC Name 6-bromo-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
SMILES BrC1=CC=C2NC(=O)OC(=O)C2=C1

Specifications

Melting Point 288°C
Quantity 5 g
Formula Weight 242.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoisatoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.