Learn More
AdipoGen 5-Bromonicotinic acid
SDP
Catalog No. 502561622
Encompass_Preferred
Chemical. CAS 20826-04-4. Formula C6H4BrNO2. MW 202.01. Synthetic. Building block for synthesis. Used in the synthesis of 3-guanidinomethyl-5-iodopyridine or as a reactant in the synthesis of porphyrins with telomerase inhibiting activity. Pharmaceutical impurity, can be used as a reference compound.
Catalog No. 50-256-1622 Supplier AdipoGen Supplier No. CDXB0248G050
Chemical. CAS 20826-04-4. Formula C6H4BrNO2. MW 202.01. Synthetic. Building block for synthesis. Used in the synthesis of 3-guanidinomethyl-5-iodopyridine or as a reactant in the synthesis of porphyrins with telomerase inhibiting activity. Pharmaceutical impurity, can be used as a reference compound.