Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromonicotinic acid, 98%, Thermo Scientific™

Catalog No. AAA1358906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1358906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1358906 Supplier Thermo Scientific Chemicals Supplier No. A1358906

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20826-04-4
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00009783
InChI Key FQIUCPGDKPXSLL-UHFFFAOYSA-N
Synonym 5-bromonicotinic acid, 5-bromo nicotinic acid, 5-bromo-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-bromo, nicotinic acid, 5-bromo, 5-bromo-nicotinic acid, 3-bromo-5-pyridine carboxylic acid, 3-bromo pyridine-5-carboxylic acid, pubchem1211, acmc-209fev
PubChem CID 88707
IUPAC Name 5-bromopyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(Br)=C1

Specifications

Melting Point 182°C to 186°C
Quantity 5 g
Beilstein 115854
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 5-Bromonicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 244-065-5
RTECSNumber QT0868000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.