Learn More
Medchemexpress LLC 5-bromopentanoic acid | 2067-33-6 | MFCD00004414 | >97.0% | 181.03 g·mol⁻¹ | C5H9BrO2 | 100 G
SDP
Catalog No. 5000303175
Encompass_Preferred
5-bromopentanoic acid is an organobromine carboxylic acid used as a building block in organic synthesis and medicinal chemistry research. It is supplied as a solid reagent (C5H9BrO2, Mw 181.03 g·mol⁻1) and is employed to introduce a bromopentanoic moiety in synthetic routes, including preparation of inhibitors for aminoglycoside-resistant bacteria. For research use only.
  • Versatile synthetic intermediate for alkylation and acylation reactions.
  • Introduces a bromopentanoic handle for further functionalization.
  • Solid form simplifies storage and handling.
  • Commonly available in gram-scale packages suitable for laboratory synthesis.
  • Characterized by CAS 2067-33-6, formula C5H9BrO2, and Mw 181.03 g·mol⁻1.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-003-03175 Supplier Medchemexpress LLC Supplier No. HYW016456100G
5-bromopentanoic acid is an organobromine carboxylic acid used as a building block in organic synthesis and medicinal chemistry research. It is supplied as a solid reagent (C5H9BrO2, Mw 181.03 g·mol⁻1) and is employed to introduce a bromopentanoic moiety in synthetic routes, including preparation of inhibitors for aminoglycoside-resistant bacteria. For research use only.
  • Versatile synthetic intermediate for alkylation and acylation reactions.
  • Introduces a bromopentanoic handle for further functionalization.
  • Solid form simplifies storage and handling.
  • Commonly available in gram-scale packages suitable for laboratory synthesis.
  • Characterized by CAS 2067-33-6, formula C5H9BrO2, and Mw 181.03 g·mol⁻1.
  • Supplied for research use only; not for human or clinical use.