Learn More
Medchemexpress LLC 5-bromopentanoic acid | 2067-33-6 | MFCD00004414 | 97.0% | 181.03 g·mol⁻¹ | C5H9BrO2 | 500 G
SDP
Catalog No. 5000301792
Encompass_Preferred
5-Bromopentanoic acid is a brominated carboxylic acid used as a chemical intermediate and building block in organic synthesis. It is commonly used in medicinal chemistry research for the preparation of derivatives and inhibitors and is supplied as a solid at laboratory-grade purity for research use only.
  • Useful intermediate for small-molecule synthesis.
  • Suitable for derivatization and coupling reactions.
  • Solid form simplifies handling and storage.
  • Typical purity 97.0% for research applications.
  • CAS number 2067-33-6 for unambiguous identification.
  • Available in multiple pack sizes, including 500 g.
Catalog No. 50-003-01792 Supplier Medchemexpress LLC Supplier No. HYW016456500G
5-Bromopentanoic acid is a brominated carboxylic acid used as a chemical intermediate and building block in organic synthesis. It is commonly used in medicinal chemistry research for the preparation of derivatives and inhibitors and is supplied as a solid at laboratory-grade purity for research use only.
  • Useful intermediate for small-molecule synthesis.
  • Suitable for derivatization and coupling reactions.
  • Solid form simplifies handling and storage.
  • Typical purity 97.0% for research applications.
  • CAS number 2067-33-6 for unambiguous identification.
  • Available in multiple pack sizes, including 500 g.