Learn More
Cayman Chemical 5-BromopentnoIc AcId-d4 1mg
SDP
Catalog No. 502990314
Encompass_Preferred
An internal standard for the quantification of 5-bromopentanoic acid by GC- or LC-MS
Catalog No. 50-299-0314 Supplier Cayman Chemical Supplier No. 309131 MG
Add to Cart
Add to Cart
An internal standard for the quantification of 5-bromopentanoic acid by GC- or LC-MS