Learn More
Cayman Chemical 5-BromopentnoIc AcId-d4 5mg
SDP
Catalog No. 502989911
Encompass_Preferred
An internal standard for the quantification of 5-bromopentanoic acid by GC- or LC-MS
Catalog No. 50-298-9911 Supplier Cayman Chemical Supplier No. 309135 MG
An internal standard for the quantification of 5-bromopentanoic acid by GC- or LC-MS