Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromophthalide | 64169-34-2 | MFCD01797360 | 213.03 | 50 G
SDP

Catalog No. 5000447190
Encompass_Preferred

5-Bromophthalide is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only and not for medical applications.

  • Biochemical reagent for life science research.
  • Solid appearance, white to light yellow color.
  • Molecular formula: C8H5BrO2.
  • Soluble in DMSO (25 mg/mL), requiring sonication.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-47190 Supplier Medchemexpress LLC Supplier No. HYW00766750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromophthalide is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only and not for medical applications.

  • Biochemical reagent for life science research.
  • Solid appearance, white to light yellow color.
  • Molecular formula: C8H5BrO2.
  • Soluble in DMSO (25 mg/mL), requiring sonication.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.