Learn More
Sigma Aldrich 5-Bromopyridine-3-acetonitrile
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Glutaconic acid, also known as 2-pentenedioic acid, is an unsaturated dicarboxylic acid
- It is formed as one of the degradation products during the partial wet oxidation (pwo) of alkali lignin
- The analysis of its crystal structure indicates that the compound exists predominantly in the trans-conformation
- The geometric bond lengths and bond angles of glutaconic acid have been obtained using hartree-fock (hf), density functional calculations and IR spectral data.glutaconic acid has been used in the preparation of coa-substrate glutaconyl-coa by reacting with acetyl-coa
- It may be used in the preparation of 6-chloro-2(2h)-pyranone by reacting with phosphorus pentachloride (pcl5)
Specifications
Specifications
| CAS | 3989-18-2 |
| Quantity | 5 g |
| MDL Number | MFCD06657722 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.