Learn More
Medchemexpress LLC 5-Bromopyrimidine | 4595-59-9 | 158.98 | 25 G
SDP
Catalog No. 5000448174
Encompass_Preferred
5-Bromopyrimidine is a biochemical reagent designed for life science research. It can be utilized as a biological material or organic compound and is intended for research use only.
  • Used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives.
  • Supports palladium-catalyzed aerobic and ligand-free Suzuki reaction.
Catalog No. 50-004-48174 Supplier Medchemexpress LLC Supplier No. 25GHYW008017
5-Bromopyrimidine is a biochemical reagent designed for life science research. It can be utilized as a biological material or organic compound and is intended for research use only.
  • Used in the synthesis of N-heteroaryl substituted 9-arylcarbazolyl derivatives.
  • Supports palladium-catalyzed aerobic and ligand-free Suzuki reaction.