Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Bromoquinoline | 4964-71-0 | 208.06 | 10 G
SDP

Catalog No. 5000445829
Encompass_Preferred

5-Bromoquinoline is a biochemical reagent and organic compound used in life science research and for the manufacture of substances. It is intended for laboratory use only.

  • Biochemical reagent for life science research
  • Used in the manufacture of substances
  • Solid appearance, white to light brown color
  • Purity of 98.1%
  • Molecular weight of 208.06
  • Stable under recommended storage conditions
  • Melting point 50-52°C
  • Boiling point 295.9°C

Catalog No. 50-004-45829 Supplier Medchemexpress LLC Supplier No. HYW00255910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Bromoquinoline is a biochemical reagent and organic compound used in life science research and for the manufacture of substances. It is intended for laboratory use only.

  • Biochemical reagent for life science research
  • Used in the manufacture of substances
  • Solid appearance, white to light brown color
  • Purity of 98.1%
  • Molecular weight of 208.06
  • Stable under recommended storage conditions
  • Melting point 50-52°C
  • Boiling point 295.9°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.