Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromosalicylic acid, 98%, Thermo Scientific™

Catalog No. AAB2033822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2033822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2033822 Supplier Thermo Scientific Chemicals Supplier No. B2033822

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromosalicylic acid is used as organic synthesis. The cross-coupling reaction of 5-bromosalicylic acid with alkyl and aryl Grignard reagents provides a navel efficient synthesis of substituted salicylic acids.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-55-4
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00002455
InChI Key IEJOONSLOGAXNO-UHFFFAOYSA-N
Synonym 5-bromosalicylic acid, benzoic acid, 5-bromo-2-hydroxy, salicylic acid, 5-bromo, 2-hydroxy-5-bromobenzoic acid, benzoic acid, 3-bromo-6-hydroxy, 5-bromo-2-hydroxy-benzoic acid, 5-bromo-2-hydroxybenzoic acid, 5-bromsalicylic acid, 5-bromosalicyclic acid, acmc-209r1n
PubChem CID 6972
IUPAC Name 5-bromo-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Br)=CC=C1O

Specifications

Melting Point 166°C to 170°C
Quantity 100 g
Beilstein 2209121
Solubility Information Soluble in water.
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 5-Bromosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-917-0
RTECSNumber VO1850000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.