Learn More
Accela Chembio Inc 5-bromothiazole | 5g | 3034-55-7 | MFCD07787394 | 95+% | D: 1.835 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt
Catalog No. 50250702
Encompass_Preferred
5-bromothiazole | 5g | 3034-55-7 | MFCD07787394 | 95+% | D: 1.835 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt
Catalog No. 50-250-702 Supplier Accela Chembio Inc Supplier No. SY0051525G
5-bromothiazole | 5g | 3034-55-7 | MFCD07787394 | 95+% | D: 1.835 | Shelf Life: 540 Days | Light Sensitive/nitrogen Or Argon/store In Freezer(-20) Upon Receipt