Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromotryptamine hydrochloride, 98%, Thermo Scientific™

Catalog No. aal19418md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19418MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19418MD Supplier Thermo Scientific Chemicals Supplier No. L19418MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromotryptamine hydrochloride is used as pharmaceutical intermediates.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with light and oxidizing agents. Store in the dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81868-12-4
Molecular Formula C10H12BrClN2
Molecular Weight (g/mol) 275.574
MDL Number MFCD00152022
InChI Key QUPXCOYSKSUJPX-UHFFFAOYSA-N
Synonym 2-5-bromo-1h-indol-3-yl ethanamine hydrochloride, 5-bromotryptamine hydrochloride, 2-5-bromo-1h-indol-3-yl-ethylamine hydrochloride, 1h-indole-3-ethanamine, 5-bromo-, monohydrochloride, 5-bromotryptamine hcl, 2-5-bromo-1h-indol-3-yl ethanaminehydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine hydrochloride, 2-5-bromo-1h-indol-3-yl ethan-1-amine-hydrogen chloride 1/1
PubChem CID 13241175
IUPAC Name 2-(5-bromo-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC2=C(C=C1Br)C(=CN2)CCN.Cl

Specifications

Melting Point ∽274°C (decomposition)
Quantity 250 mg
Beilstein 3728224
Sensitivity Light sensitive
Formula Weight 275.58
Percent Purity 98%
Chemical Name or Material 5-Bromotryptamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.