Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Bromouracil

Catalog No. 85247325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-247-325G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 85-247-325G Supplier Sigma Aldrich Supplier No. 85247325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phthalic acid mono-2-ethylhexyl ester can be used as a precursor:
  • To synthesize dioctyl phthalate, a widely used plasticizer, via esterification reaction with 2-ethylhexanol
  • To prepare a fluorescent probe, 2-[[(3α,6α-dihydroxy-3-oxospiro[isobenzofuran-1(3h),9α-[9h]xanthen]-5-yl)amino]carbonyl] 2-ethylhexyl benzoic acid ester, to detect toxic metabolite mono-2-ethylhexyl phthalate (mehp)

Specifications

CAS 51-20-7
Melting Point >300°C (lit.)
Molecular Formula C4H3BrN2O2
Linear Formula C4H3BrN2O2
MDL Number MFCD00006017
Quantity 500 mg
Synonym 5-Bromo-2,4-dihydroxypyrimidine
Molecular Weight (g/mol) 190.98
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.