Learn More
Cayman Chemical 5-Bromouridine, Purity: ≥98%, CAS: 957-75-5, 500 mg
SDP
Catalog No. 502990186
Encompass_Preferred
A brominated uridine analog; can be incorporated into RNA and subsequently detected by antibodies against bromodeoxyuridine; decreases the viability of HL-60 and MOLT-4 cells (LC50s = 10 and 20 μM, respectively); induces apoptosis and halts the cell cycle at the S phase in HL-60 cells; photoreactive; UV irradiation has been used to cross-link RNA containing 5-bromouridine to proteins in the study of RNA-protein interactions; can be incorporated into RNA using 5-bromouridine 5’-triphosphate
Catalog No. 50-299-0186 Supplier Cayman Chemical Supplier No. 30457500 MG
A brominated uridine analog; can be incorporated into RNA and subsequently detected by antibodies against bromodeoxyuridine; decreases the viability of HL-60 and MOLT-4 cells (LC50s = 10 and 20 μM, respectively); induces apoptosis and halts the cell cycle at the S phase in HL-60 cells; photoreactive; UV irradiation has been used to cross-link RNA containing 5-bromouridine to proteins in the study of RNA-protein interactions; can be incorporated into RNA using 5-bromouridine 5’-triphosphate