Learn More
Medchemexpress LLC 5-bromouridine 5'-triphosphate sodium salt | 161848-60-8 | MFCD00274403 | 98.0% | 563.04 g/mol | C9H14BrN2O15P3.xNa | 1 MG
SDP
Catalog No. 5000248925
Encompass_Preferred
5-BrUTP sodium salt is a brominated uridine triphosphate used as a nucleotide analog to label RNA in transcription assays. It can be enzymatically incorporated into nascent RNA during in vitro transcription or cellular labeling, and incorporated BrUTP residues are detected immunologically to quantify or visualize newly synthesized RNA.
  • Enables labeling of newly synthesized RNA for transcription analysis.
  • Detectable by anti-BrdU antibodies for immunodetection.
  • Brominated uridine analog compatible with enzymatic incorporation during transcription.
  • Sodium salt form offers improved aqueous solubility for biochemical workflows.
  • High purity (98.0%) suitable for sensitive biochemical assays.
  • Available in multiple pack sizes to support small- and large-scale experiments.
Catalog No. 50-002-48925 Supplier Medchemexpress LLC Supplier No. HY1018861MG
5-BrUTP sodium salt is a brominated uridine triphosphate used as a nucleotide analog to label RNA in transcription assays. It can be enzymatically incorporated into nascent RNA during in vitro transcription or cellular labeling, and incorporated BrUTP residues are detected immunologically to quantify or visualize newly synthesized RNA.
  • Enables labeling of newly synthesized RNA for transcription analysis.
  • Detectable by anti-BrdU antibodies for immunodetection.
  • Brominated uridine analog compatible with enzymatic incorporation during transcription.
  • Sodium salt form offers improved aqueous solubility for biochemical workflows.
  • High purity (98.0%) suitable for sensitive biochemical assays.
  • Available in multiple pack sizes to support small- and large-scale experiments.