Learn More
Indofine Chemical 5-Bromovanillin, 97%, 25 Gm
SDP
Catalog No. 50908312
Encompass_Preferred
5-BROMOVANILLIN 25 GM, Rare Organics & Biochemicals, 5-Bromo-4-hydroxy-3-methoxybenzaldehyde: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde, MW: 231.05, 97%, 2973-76-4, MFCD00006940
Catalog No. 50-908-312 Supplier Indofine Chemical Supplier No. B006 25 GM
5-BROMOVANILLIN 25 GM, Rare Organics & Biochemicals, 5-Bromo-4-hydroxy-3-methoxybenzaldehyde: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde, MW: 231.05, 97%, 2973-76-4, MFCD00006940