Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-carboxy-2'-deoxycytidine | 1210427-59-0 | 98.1% | 10 MG
SDP

Catalog No. 5000295047
Encompass_Preferred

5-Carboxy-2′-deoxycytidine is a deoxyribonucleoside derivative and the primary metabolite of trifluridine. Supplied as a high-purity research reagent, it is intended for studies of nucleoside metabolism, drug metabolism, and biochemical assays. The compound is provided in small milligram quantities suitable for analytical and preparative work.

  • Metabolite of trifluridine useful for drug metabolism studies.
  • High purity: 98.1% (as supplied).
  • Molecular formula: C10H13N3O6, molecular weight: 271.23 g/mol.
  • Supplied in milligram quantities suitable for research assays.
  • CAS number 1210427-59-0 for unambiguous identification.

Catalog No. 50-002-95047 Supplier Medchemexpress LLC Supplier No. HY17325810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Carboxy-2′-deoxycytidine is a deoxyribonucleoside derivative and the primary metabolite of trifluridine. Supplied as a high-purity research reagent, it is intended for studies of nucleoside metabolism, drug metabolism, and biochemical assays. The compound is provided in small milligram quantities suitable for analytical and preparative work.

  • Metabolite of trifluridine useful for drug metabolism studies.
  • High purity: 98.1% (as supplied).
  • Molecular formula: C10H13N3O6, molecular weight: 271.23 g/mol.
  • Supplied in milligram quantities suitable for research assays.
  • CAS number 1210427-59-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.