Learn More
Medchemexpress LLC 5-carboxy-2'-deoxyuridine | 14599-46-3 | 272.21 | C10H12N2O7 | 10 MG
SDP
Catalog No. 5000281465
Encompass_Preferred
5-Carboxy-2'-deoxyuridine is a pyrimidine nucleoside metabolite of trifluridine provided as a powder for research and analytical applications. It is intended for preparation of stock solutions and for use as an analytical standard in studies of nucleotide metabolism and drug metabolite profiling.
  • Supplied as a powder suitable for preparing concentrated stock solutions.
  • Intended for use as an analytical standard and research reagent.
  • Well characterized: CAS 14599-46-3; molecular weight 272.21.
  • Stable under recommended storage conditions for long-term retention.
  • High solubility in DMSO (125 mg/mL) for convenient formulation.
Catalog No. 50-002-81465 Supplier Medchemexpress LLC Supplier No. HYW42477910MG
5-Carboxy-2'-deoxyuridine is a pyrimidine nucleoside metabolite of trifluridine provided as a powder for research and analytical applications. It is intended for preparation of stock solutions and for use as an analytical standard in studies of nucleotide metabolism and drug metabolite profiling.
  • Supplied as a powder suitable for preparing concentrated stock solutions.
  • Intended for use as an analytical standard and research reagent.
  • Well characterized: CAS 14599-46-3; molecular weight 272.21.
  • Stable under recommended storage conditions for long-term retention.
  • High solubility in DMSO (125 mg/mL) for convenient formulation.