Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-carboxy-2'-deoxyuridine | 14599-46-3 | 272.21 g/mol | C10H12N2O7 | 25 MG
SDP

Catalog No. 5000281466
Encompass_Preferred

5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a metabolite of trifluridine supplied as a solid for research and analytical use. It is a methyl-oxidation product of thymidine and is intended for laboratory and analytical applications only.

  • Molecular formula: C10H12N2O7.
  • Molecular weight: 272.21 g/mol.
  • CAS number: 14599-46-3.
  • Physical form: powder for research use.
  • Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C recommended.
  • Applications: analytical standard and metabolite studies.

Catalog No. 50-002-81466 Supplier Medchemexpress LLC Supplier No. HYW42477925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a metabolite of trifluridine supplied as a solid for research and analytical use. It is a methyl-oxidation product of thymidine and is intended for laboratory and analytical applications only.

  • Molecular formula: C10H12N2O7.
  • Molecular weight: 272.21 g/mol.
  • CAS number: 14599-46-3.
  • Physical form: powder for research use.
  • Storage: powder -20°C (long term) or 4°C (short term); in solvent -80°C recommended.
  • Applications: analytical standard and metabolite studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.