Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-carboxy-2'-deoxyuridine | 14599-46-3 | 99.0% | 272.21 g/mol | C10H12N2O7 | 100 MG
SDP

Catalog No. 5000280723
Encompass_Preferred

5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a known metabolite of the antiviral trifluridine. It is supplied as an analytical reference standard for research applications, including metabolite identification and method development for chromatographic and mass spectrometric assays. Handle and store according to recommended conditions and consult certificate of analysis for purity and lot-specific data.

  • Used as an analytical reference standard for metabolite identification.
  • Compatible with LC-MS and other mass spectrometry workflows.
  • Molecular weight 272.21 g/mol; formula C10H12N2O7.
  • Recommended storage: powder at -20°C for long-term stability.
  • Provided in milligram-scale quantities appropriate for analytical use.

Catalog No. 50-002-80723 Supplier Medchemexpress LLC Supplier No. HYW424779100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a known metabolite of the antiviral trifluridine. It is supplied as an analytical reference standard for research applications, including metabolite identification and method development for chromatographic and mass spectrometric assays. Handle and store according to recommended conditions and consult certificate of analysis for purity and lot-specific data.

  • Used as an analytical reference standard for metabolite identification.
  • Compatible with LC-MS and other mass spectrometry workflows.
  • Molecular weight 272.21 g/mol; formula C10H12N2O7.
  • Recommended storage: powder at -20°C for long-term stability.
  • Provided in milligram-scale quantities appropriate for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.