Learn More
Medchemexpress LLC 5-carboxy-2'-deoxyuridine | 14599-46-3 | MFCD22495153 | 272.21 g·mol⁻¹ | C10H12N2O7 | 1 MG
SDP
Catalog No. 5000280675
Encompass_Preferred
5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a known metabolite of the antiviral trifluridine. It is the methyl-oxidation product of thymidine and is supplied as an analytical standard for research applications, particularly studies of nucleoside metabolism and drug biotransformation.
  • Provides a validated reference for metabolite and biotransformation studies.
  • Chemical identity: CAS 14599-46-3; formula C10H12N2O7; MW 272.21 g·mol⁻¹.
  • Available in small quantities suitable for analytical workflows (1 mg, 5 mg, 10 mg).
  • Intended for laboratory research and analytical use.
Catalog No. 50-002-80675 Supplier Medchemexpress LLC Supplier No. HYW4247791MG
5-Carboxy-2'-deoxyuridine is a deoxyuridine derivative and a known metabolite of the antiviral trifluridine. It is the methyl-oxidation product of thymidine and is supplied as an analytical standard for research applications, particularly studies of nucleoside metabolism and drug biotransformation.
  • Provides a validated reference for metabolite and biotransformation studies.
  • Chemical identity: CAS 14599-46-3; formula C10H12N2O7; MW 272.21 g·mol⁻¹.
  • Available in small quantities suitable for analytical workflows (1 mg, 5 mg, 10 mg).
  • Intended for laboratory research and analytical use.