Learn More
Medchemexpress LLC 5-carboxy-2-thiouracil | 23945-50-8 | 172.16 | C5H4N2O3S | 1 G
SDP
Catalog No. 5000209974
Encompass_Preferred
5-Carboxy-2-thiouracil is a small-molecule biochemical reagent reported to have antitumor activity; it can chelate metal ions and is used in biochemical assays and research.
  • Reported antitumor activity observed in biochemical studies.
  • Capable of forming complexes with metal ions (chelating properties).
  • Supplied as a solid for use in research and assay development.
  • Commonly used in biochemical assays and metal-coordination studies.
Catalog No. 50-002-09974 Supplier Medchemexpress LLC Supplier No. HYW0527501G
Add to Cart
Add to Cart
5-Carboxy-2-thiouracil is a small-molecule biochemical reagent reported to have antitumor activity; it can chelate metal ions and is used in biochemical assays and research.
  • Reported antitumor activity observed in biochemical studies.
  • Capable of forming complexes with metal ions (chelating properties).
  • Supplied as a solid for use in research and assay development.
  • Commonly used in biochemical assays and metal-coordination studies.