Learn More
AdipoGen 5-Carboxy-DCF DA
SDP
Supplier:  AdipoGen CDXC0333M250
Encompass_Preferred
Chemical. CAS 144489-09-8. Formula C25H14Cl2O9. MW 529.28. Synthetic. Non-activated, amine-reactive fluorescent probe. Single 5-isomer of CDCFDA that can be used in biomolecules conjugation. CDCFDA is an amine–reactive fluorescein diacetate FDA derivative used to prepare a variety of FDA conjugates. CDCFDA is a non-fluorescent molecule that diffuse into cells and is hydrolyzed by intracellular non-specific esterases, yielding a fluorescent probe. The probe accumulates exclusively in viable cells with intact cell membranes. CFCFDA does not stain dead cells. CDCFDA labeling can be used for monitoring cells by flow cytometry or fluorescence microscopy. The probe is also frequently used to monitor reactive oxygen species ROS generation in live cells and intracellular pH levels. Spectral data lambdaex=504nm, lambdaem=529nm.
Catalog No. 50-256-1379