Learn More
AdipoGen 5-Carboxy-DCFDA (100 mg)
SDP
Catalog No. 501490318
Encompass_Preferred
5-Carboxy-DCFDA (100 mg), AdipoGen Life Sciences. Chemical. CAS: 144489-09-8. Formula: C25H14Cl2O9. MW: 529.28. Synthetic. Non-activated, amine-reactive fluorescent probe. Single 5-isomer of CDCFDA that can be used in biomolecules conjugation. CDCFDA is an amine–reactive fluorescein diacetate (FDA) derivative used to prepare a variety of FDA conjugates. CDCFDA is a non-fluorescent molecule that diffuse into cells and is hydrolyzed by intracellular non-specific esterases, yielding a fluorescent probe. The probe accumulates exclusively in viable cells with intact cell membranes. CFCFDA does not stain dead cells. CDCFDA labeling can be used for monitoring cells by flow cytometry or fluorescence microscopy. The probe is also frequently used to monitor reactive oxygen species (ROS) generation in live cells and intracellular pH levels. Spectral data: lambdaex=504nm, lambdaem=529nm.
Catalog No. 50-149-0318 Supplier AdipoGen Supplier No. CDXC0333M100
5-Carboxy-DCFDA (100 mg), AdipoGen Life Sciences. Chemical. CAS: 144489-09-8. Formula: C25H14Cl2O9. MW: 529.28. Synthetic. Non-activated, amine-reactive fluorescent probe. Single 5-isomer of CDCFDA that can be used in biomolecules conjugation. CDCFDA is an amine–reactive fluorescein diacetate (FDA) derivative used to prepare a variety of FDA conjugates. CDCFDA is a non-fluorescent molecule that diffuse into cells and is hydrolyzed by intracellular non-specific esterases, yielding a fluorescent probe. The probe accumulates exclusively in viable cells with intact cell membranes. CFCFDA does not stain dead cells. CDCFDA labeling can be used for monitoring cells by flow cytometry or fluorescence microscopy. The probe is also frequently used to monitor reactive oxygen species (ROS) generation in live cells and intracellular pH levels. Spectral data: lambdaex=504nm, lambdaem=529nm.