Learn More
Bachem 5-Carboxy-fluorescein
SDP
Catalog No. 50260666
Encompass_Preferred
5-Carboxy-fluorescein, 76823-03-5, C21H12O7, Mr 376.32, 500mg. 5-FAM, 4-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-isophthalic acid. Widely used green-fluorescent labeling dye.
Catalog No. 50-260-666 Supplier Bachem Supplier No. 4045141.0500
5-Carboxy-fluorescein, 76823-03-5, C21H12O7, Mr 376.32, 500mg. 5-FAM, 4-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-isophthalic acid. Widely used green-fluorescent labeling dye.