Learn More
Cayman Chemical 5-Carboxyfluorescein, Purity: ≥98%, CAS: 76823-03-5, 100 mg
SDP
Catalog No. 502984569
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-4569 Supplier Cayman Chemical Supplier No. 19581100 MG
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively