Learn More
Cayman Chemical 5-CarboxyfluoresceIn 1g
SDP
Catalog No. 502984433
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-4433 Supplier Cayman Chemical Supplier No. 195811 G
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively