Learn More
Cayman Chemical 5-Carboxyfluorescein, Purity: ≥98%, CAS: 76823-03-5, 25 mg
SDP
Catalog No. 502983260
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-3260 Supplier Cayman Chemical Supplier No. 1958125 MG
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively