Learn More
Cayman Chemical 5-CarboxyfluoresceIn 50mg
SDP
Catalog No. 502981976
Encompass_Preferred
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively
Catalog No. 50-298-1976 Supplier Cayman Chemical Supplier No. 1958150 MG
Add to Cart
Add to Cart
A single isomer derivative of 5(6)-carboxyfluorescein that can be used to fluorescently label biomolecules; excitation/emission maxima of 492 and 518 nm, respectively