Learn More
Apexbio Technology LLC 5-Carboxyfluorescein diacetate N-succinimidyl ester 150206-05-6 10mg
SDP
Catalog No. 5000095982
Encompass_Preferred
5-Carboxyfluorescein diacetate N-succinimidyl ester (CAS 150206-05-6) is a fluorescent cell-permeable dye It is designed to enable stable cell labeling by facilitating intracellular conversion and covalent attachment to cellular amines 5-Carboxyfluorescein diacetate N-succinimidyl ester exerts its activity through membrane permeation intracellular esterase-mediated cleavage of acetate groups and subsequent covalent coupling to intracellular targets resulting in persistent fluorescence through cell division Based on these properties 5-Carboxyfluorescein diacetate N-succinimidyl ester holds research potential in tracking lymphocyte migration analyzing cell proliferation and investigating cell dynamics and interactions in various in vitro and in vivo experimental models
Catalog No. 50-000-95982 Supplier Apexbio Technology LLC Supplier No. A896810
5-Carboxyfluorescein diacetate N-succinimidyl ester (CAS 150206-05-6) is a fluorescent cell-permeable dye It is designed to enable stable cell labeling by facilitating intracellular conversion and covalent attachment to cellular amines 5-Carboxyfluorescein diacetate N-succinimidyl ester exerts its activity through membrane permeation intracellular esterase-mediated cleavage of acetate groups and subsequent covalent coupling to intracellular targets resulting in persistent fluorescence through cell division Based on these properties 5-Carboxyfluorescein diacetate N-succinimidyl ester holds research potential in tracking lymphocyte migration analyzing cell proliferation and investigating cell dynamics and interactions in various in vitro and in vivo experimental models