Learn More
Medchemexpress LLC 5-Carboxyfluorescein diacetate N-succinimidyl ester | 150206-05-6 | 99.3% | 100 MG
SDP
Catalog No. 5000410660
Encompass_Preferred
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell permeable dye that labels cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation.
  • Cell permeable dye
  • Labels cells by covalently binding to intracellular molecules
  • Used to track lymphocyte migration and proliferation
  • Excitation (Ex): 492 nm
  • Emission (Em): 517 nm
  • Solid, white to off-white appearance
Catalog No. 50-004-10660 Supplier Medchemexpress LLC Supplier No. HYD0056100MG
Add to Cart
Add to Cart
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell permeable dye that labels cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation.
  • Cell permeable dye
  • Labels cells by covalently binding to intracellular molecules
  • Used to track lymphocyte migration and proliferation
  • Excitation (Ex): 492 nm
  • Emission (Em): 517 nm
  • Solid, white to off-white appearance