Learn More
Medchemexpress LLC 5-Carboxyfluorescein diacetate N-succinimidyl ester | 150206-05-6 | 99.3% | 25 MG
SDP
Catalog No. 5000411275
Encompass_Preferred
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell-permeable dye that can label cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation.
  • Cell permeable dye.
  • Labels cells by covalently binding to intracellular molecules.
  • Used to track lymphocyte migration and proliferation.
  • Excitation at 482 nm.
  • Emission at 525 nm.
Catalog No. 50-004-11275 Supplier Medchemexpress LLC Supplier No. 25MGHYD0056
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell-permeable dye that can label cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation.
  • Cell permeable dye.
  • Labels cells by covalently binding to intracellular molecules.
  • Used to track lymphocyte migration and proliferation.
  • Excitation at 482 nm.
  • Emission at 525 nm.