Learn More
Medchemexpress LLC 5-Carboxyfluorescein diacetate N-succinimidyl ester | 150206-05-6 | 99.3% | 50 MG
SDP
Catalog No. 5000410661
Encompass_Preferred
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell permeable dye that labels cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation, and is suitable for laboratory research.
  • Cell permeable dye.
  • Covalently binds to intracellular molecules.
  • Used to track lymphocyte migration and proliferation.
  • Excitation (Ex): 482 nm.
  • Emission (Em): 525 nm.
Catalog No. 50-004-10661 Supplier Medchemexpress LLC Supplier No. HYD005650MG
Add to Cart
Add to Cart
5-Carboxyfluorescein diacetate N-succinimidyl ester is a cell permeable dye that labels cells by covalently binding to intracellular molecules. It is used to track lymphocyte migration and proliferation, and is suitable for laboratory research.
  • Cell permeable dye.
  • Covalently binds to intracellular molecules.
  • Used to track lymphocyte migration and proliferation.
  • Excitation (Ex): 482 nm.
  • Emission (Em): 525 nm.