Learn More
Alkali Scientific 5" Cardboard Freezer Boxes with Drain Holes Water and Ice Resistant Coating, w/ 64 Dividers for 6mL 13 x100mm Tubes, 50/Case
SDP
Catalog No. 5000662530
Encompass_Preferred
• Choose optional slot/cell dividers • Dividers also sold separately, item CD • Temperature Range: 196 to +121 • Drain holes in bottom corners • Writing area on the lid • Keeps tubes in an upright position for easy sorting Alkali Scientific™ 5" cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box 5 3/16x 5 3/16 x 4 15/16″ ( 148 x 148 x 125mm)
Catalog No. 50-006-62530 Supplier Alkali Scientific Supplier No. FB5C64C50
• Choose optional slot/cell dividers • Dividers also sold separately, item CD • Temperature Range: 196 to +121 • Drain holes in bottom corners • Writing area on the lid • Keeps tubes in an upright position for easy sorting Alkali Scientific™ 5" cardboard freezer box are ideal for any chemical laboratories, medicinal facilities or healthcare units for a wide range of purposes such as scientific research and development. Cardboard Freezer Box 5 3/16x 5 3/16 x 4 15/16″ ( 148 x 148 x 125mm)