Learn More
Abcam 5-CFDA (5-Carbo x yfluorescein Diacetate), Fluorogenic non-selective esterase substrate, 25MG
Catalog No. 502705355
Encompass_Preferred
MW 460.4 Da, Purity >95%. Hydrophobic, cell-permeable fluorogenic non-selective esterase substrate. Becomes hydrolyzed to hydrophilic strongly fluorescent 5-Carboxyfluorescein (ab145293) intracellularly. Measures intracellular pH. 5-CFDA N-succinimidyl ester (ab145292) precursor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5355 Supplier Abcam Supplier No. AB14525625MG
MW 460.4 Da, Purity >95%. Hydrophobic, cell-permeable fluorogenic non-selective esterase substrate. Becomes hydrolyzed to hydrophilic strongly fluorescent 5-Carboxyfluorescein (ab145293) intracellularly. Measures intracellular pH. 5-CFDA N-succinimidyl ester (ab145292) precursor.
The product is subject to the following: Abcam Restricted Use Statement