Learn More
AdipoGen 5-CFDA ethanedioic-S-PhMeEster
SDP
Supplier:  AdipoGen CDXC0051M010
Encompass_Preferred
Chemical. CAS 690958-19-1. Formula C34H25NO9S. MW 623.6. Synthetic. Cell permeable fluorescent probe. Used for site-specific labeling of N-terminal cysteine proteins with the small molecule probes.
Catalog No. 50-256-1511