Learn More
AdipoGen 5-CFDA ethanedioic-S-PhMeEster
SDP
Catalog No. 502561510
Encompass_Preferred
Chemical. CAS 690958-19-1. Formula C34H25NO9S. MW 623.6. Synthetic. Cell permeable fluorescent probe. Used for site-specific labeling of N-terminal cysteine proteins with the small molecule probes.
Catalog No. 50-256-1510 Supplier AdipoGen Supplier No. CDXC0051M050
Chemical. CAS 690958-19-1. Formula C34H25NO9S. MW 623.6. Synthetic. Cell permeable fluorescent probe. Used for site-specific labeling of N-terminal cysteine proteins with the small molecule probes.