Learn More
Biotium 5-CFDA (5-Carboxyfluorescein Diacetate,
Catalog No. 501964708
Encompass_Preferred
5-CFDA is membrane-permeant and thus can be loaded into cells via incubation. Once inside cells, 5-CFDA is hydrolyzed by intracellular esterases to 5-carboxyfluorescein. Properties: Off-white solid soluble in DMSO; Store desiccated at 4°C; C25H16O9; MW: 460; [79955-27-4].
Catalog No. 50-196-4708 Supplier Biotium Supplier No. 51018
5-CFDA is membrane-permeant and thus can be loaded into cells via incubation. Once inside cells, 5-CFDA is hydrolyzed by intracellular esterases to 5-carboxyfluorescein. Properties: Off-white solid soluble in DMSO; Store desiccated at 4°C; C25H16O9; MW: 460; [79955-27-4].