Learn More
Checkers 5-Channel AMS Accessibility Ramps and Rails Kit 3
Catalog No. 19088409 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Black on Yellow
Catalog No. Color
19-088-409 Black on Yellow
1 options
Catalog No. 19-088-409 Supplier Checkers Supplier No. WSA125KIT3HS
Only null left
Add to Cart
Add to Cart
5-Channel Yellow Jacket™ AMS™ (Advanced Modular System) Accessibility Ramps Kit 3 features a gradual slope and slip-resistant surface to ensure a safe crossing point over cable protectors for carts, forklifts, vehicles with small wheels, and heavy trucks.
Kit 3 - Includes:
  • 2 Ramps
  • 1 AMS™ Center Sections
Specifications
Color Black on Yellow
Material HDPE
Product Type Cable Protector Kit