Learn More
Andwin Scientific 5-CHANNEL TIMER CERTIFIED
SDP
Catalog No. 50949488
Encompass_Preferred
The 5 Channel Timer features 4 independent channels, each of which counts up or down for any interval from 1 second to 100 hours. It has an extra large display that shows the channels being used. This product can automatically start to count up to indicate time elapsed after count down reaches zero. The timer has a stand, pocket clip, and magnet, allowing this to be placed almost anywhere. It measures 2 7/8 x2 1/2 x 9/16".
Catalog No. 50-949-488 Supplier Andwin Scientific Supplier No. 7260002
The 5 Channel Timer features 4 independent channels, each of which counts up or down for any interval from 1 second to 100 hours. It has an extra large display that shows the channels being used. This product can automatically start to count up to indicate time elapsed after count down reaches zero. The timer has a stand, pocket clip, and magnet, allowing this to be placed almost anywhere. It measures 2 7/8 x2 1/2 x 9/16".