Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. C07861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07861G 1 g
C0786100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07861G Supplier TCI America Supplier No. C07861G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4199-89-7
Molecular Formula C12H7ClN2
Molecular Weight (g/mol) 214.65
MDL Number MFCD00004980
InChI Key XDUUQOQFSWSZSM-UHFFFAOYSA-N
PubChem CID 77865
IUPAC Name 5-chloro-1,10-phenanthroline
SMILES ClC1=CC2=CC=CN=C2C2=NC=CC=C12

Specifications

Melting Point 123°C
Quantity 1 g
Synonym 1,10-phenanthroline, 5-chloro, 5-chloropyridino 3,2-h quinoline, acmc-209jmo, 1,10-phenanthroline,5-chloro, 5-chloro-1,10 phenanthroline, 5-chloro-1,10-phenanthroline, 5-chloro-1, 10-phenanthroline
Formula Weight 214.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.