Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1-indanone 97.0+%, TCI America™
SDP

Catalog No. C16445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16445G 5 g
C16441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16445G Supplier TCI America Supplier No. C16445G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 42348-86-7
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.60
MDL Number MFCD00041456
InChI Key MEDSHTHCZIOVPU-UHFFFAOYSA-N
Synonym 5-chloro-1-indanone, 5-chloro-2,3-dihydro-1h-inden-1-one, 5-chloroindan-1-one, 5-chloro-indan-1-one, 1h-inden-1-one, 5-chloro-2,3-dihydro, 5-chloroindanone, 5-chloroindone, pubchem2250, 1-indanone, 5-chloro, 5-chloro indanone
PubChem CID 142599
IUPAC Name 5-chloro-2,3-dihydro-1H-inden-1-one
SMILES ClC1=CC=C2C(=O)CCC2=C1

Specifications

Melting Point 94°C
Quantity 5 g
Formula Weight 166.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.