Learn More
eMolecules​ 137048-96-5 | 5-Chloro-1-methyl-1h-imidazole-4-sulfonyl chloride | Combi-Blocks | MFCD09044274 | 215.050 | C4H4Cl2N2O2S | 95.000 | Cn1cnc(c1Cl)S(Cl)(=O)=O | 1g | 117571590
Supplier:  eMolecules​ 117571590
Encompass_Preferred
5-Chloro-1-methyl-1h-imidazole-4-sulfonyl chloride | Combi-Blocks | 137048-96-5 | MFCD09044274 | 215.050 | C4H4Cl2N2O2S | 95.000 | Cn1cnc(c1Cl)S(Cl)(=O)=O | 1g | 117571590
Catalog No. NC1647903