Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Chloro-1-pentyne

Catalog No. 2443765G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-437-65G 5 g
24-437-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-437-65G Supplier Sigma Aldrich Supplier No. 2443765G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 5-chloro-1-pentyne was used in the synthesis of 5-chloro-2-trimethylstannyl-1-pentene by reacting with me3sncu.me2s
  • It was also used with α-diazoesters in a copper-catalyzed synthesis of pyrazoles

Specifications

CAS 14267-92-6
Density 0.968 g/mL (at 25°C (literature))
Boiling Point 67°C to 69°C (145 mmHg)
Molecular Formula C5H7Cl
Refractive Index n20/D 1.444 (literature)
Linear Formula HC///CCH2CH2CH2Cl
MDL Number MFCD00001014
Quantity 5 g
Molecular Weight (g/mol) 102.56
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.