Learn More
Medchemexpress LLC 5-chloro-2-(2,4-dichlorophenoxy-1,2,3,4,5,6-13C6)phenol | 2726926-25-4 | MFCD00800992 | 99.5% | 295.56 g/mol | C613C6H7Cl3O2 | 1 MG
SDP
Catalog No. 5000284301
Encompass_Preferred
Triclosan-13C6 is a six-13C isotopically labeled triclosan supplied as an analytical standard for use as an internal standard in mass spectrometry and isotope-dilution workflows. It is high-purity, intended for analytical applications, and has defined storage recommendations for stability.
  • 13C6-labeled compound suitable for use as an internal standard.
  • High HPLC purity (99.51% as reported by Certificate of Analysis).
  • Provided in a 1 mg quantity appropriate for analytical workflows.
  • Storage recommendations included for neat material and solutions to preserve stability.
  • Isotopic labeling facilitates accurate quantitation in mass spectrometry assays.
Catalog No. 50-002-84301 Supplier Medchemexpress LLC Supplier No. HYW6539621MG
Triclosan-13C6 is a six-13C isotopically labeled triclosan supplied as an analytical standard for use as an internal standard in mass spectrometry and isotope-dilution workflows. It is high-purity, intended for analytical applications, and has defined storage recommendations for stability.
  • 13C6-labeled compound suitable for use as an internal standard.
  • High HPLC purity (99.51% as reported by Certificate of Analysis).
  • Provided in a 1 mg quantity appropriate for analytical workflows.
  • Storage recommendations included for neat material and solutions to preserve stability.
  • Isotopic labeling facilitates accurate quantitation in mass spectrometry assays.